Tisk…

Agaróza for electrophoresis, for electrophoresis medium electroendosmosis (EEO) (Mr = 0.16 - 0.19)

Dodavatel: Thermo Fisher Scientific

400420250 400421000 400425000
ACRO400420250EA 2320 CZK
ACRO400420250 ACRO400421000 ACRO400425000
Agaróza for electrophoresis, for electrophoresis medium electroendosmosis (EEO) (Mr = 0.16 - 0.19)
Agaróza
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS číslo: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White to off-white to cream Powder
Infrared spectrum Conforms
Melting point 86.5 to 89.5 °C (1.5 % gel)
Loss on drying ≤10 % (5 g, 110 °C, 24 h)
Gel strength ≥750 g/cm² (1 % gel)
Gel point 34.5 to 37.5 °C (1.5 % gel)
Electroendosmosis 0.16 to 0.19 (-Mr)
Ash ≤0.5 %
Sulfate (SO4) ≤0.25 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR