Tisk…

Methyl hydrogen isophthalate 97%

Dodavatel: Thermo Fisher Scientific

398230010 398230050
ACRO398230010EA 1440 CZK
ACRO398230010 ACRO398230050
Methyl hydrogen isophthalate 97%
Methyl hydrogen isophthalate
Vzorec: C₉H₈O₄
Bod tání: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00029972
CAS číslo: 1877-71-0

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 191 to 197 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR