Tisk…

N,N-Diisopropylethylamine trihydrofluoride 90%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N-Ethyl-N-isopropylpropan-2-amine trihydrofluoride

390710050 390710010
ACRO390710050EA 5080 CZK
ACRO390710050 ACRO390710010
N,N-Diisopropylethylamine trihydrofluoride 90%
N,N-Diisopropylethylamine trihydrofluoride

Appearance: Clear colorless to yellow-brown Liquid

Vzorec: C₈H₂₂F₃N
MW: 189.266 g/mol
Bod varu: 93…95 °C (4 mmHg)
Hustota: 0.965 g/cm³
Bod vzplanutí: 40.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00144880
CAS číslo: 131600-43-6
UN: 3286
ADR: 3 II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Titration with NaOH 18 to 22 % (available HF)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR