Tisk…

Cellulose, microcrystalline ∼ Ø 90 µm, extra pure

Dodavatel: Thermo Fisher Scientific

382312500 382310010 382310050 382310250
ACRO382312500EA 2420 CZK
ACRO382312500 ACRO382310010 ACRO382310050 ACRO382310250
Cellulose, microcrystalline ∼ Ø 90 µm, extra pure
Cellulose
Vzorec: (C₁₂H₂₂O₁₁)n
Bod tání: 260 - 270 °C
Hustota: 1,27 to 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081512
CAS číslo: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Loss on drying ≤1.5 %
Heavy metals ≤10 ppm
Particle size ≥70 % (> 32 µm)
Residue after ignition ≤0.05 %
pH 5 to 7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR