Tisk…

2-Aminoimidazole hemisulphate 98+%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)

368860025 368860100
ACRO368860025EA 2860 CZK
ACRO368860025 ACRO368860100
2-Aminoimidazole hemisulphate 98+%
2-Aminoimidazole hemisulphate
Vzorec: C₆H₁₂N₆O₄S
Storage Temperature: Ambient
MDL Number: MFCD00013162
CAS číslo: 1450-93-7
EINECS: 215-918-9

Objednat

Výsledky specifikačního testu

Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR