Tisk…

L(+)-Arabinóza 99% for biochemistry

Dodavatel: Thermo Fisher Scientific

365180250 365181000 365185000
ACRO365180250EA 1740 CZK
ACRO365180250 ACRO365181000 ACRO365185000
L(+)-Arabinóza 99% for biochemistry
L(+)-Arabinóza
Vzorec: C₅H₁₀O₅
Bod tání: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00006609
CAS číslo: 87-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Solubility (10 % in water) Clear colorless
HPLC ≥98.5 % (ELSD)
UV at 280 nm A: ≤0.1
Residue after ign. (sulfated) ≤0.3 %
Specific optical rotation (after 24 h)
Heavy metals (as Pb) ≤10 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR