Tisk…

(R)-2-Isopropylsuccinic acid-1-methyl ester ≥95%, ee 98%

Dodavatel: Thermo Fisher Scientific

363065000 363060050
ACRO363065000EA 6620 CZK
ACRO363065000 ACRO363060050
(R)-2-Isopropylsuccinic acid-1-methyl ester ≥95%, ee 98%
(R)-2-Isopropylsuccinic acid-1-methyl ester

CAS No.: 220498-08-8

Vzorec: C₈H₁₄O₄
MW: 174.196 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01091030
CAS číslo: 220498-08-8

Objednat

Výsledky specifikačního testu

Appearance Clear yellow Viscous liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR