Tisk…

Gallium(III) trifluoromethanesulphonate 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Gallium tris(trifluoromethanesulphonate)

343930010
ACRO343930010EA 2760 CZK
ACRO343930010
Gallium(III) trifluoromethanesulphonate 99%
Gallium(III) trifluoromethanesulphonate

Appearance: White to light beige Powder

Vzorec: C₃F₉GaO₉S₃
Hustota: 1,7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD02093898
CAS číslo: 74974-60-0
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to light beige Powder
Infrared spectrum Conforms
Assay ≥98.5 % (ex F)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR