Tisk…

Chlorodimethylvinylsilane 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Dimethylvinylchlorosilane, Vinyldimethylchlorosilane

336730050 336730250
ACRO336730050EA 1080 CZK
ACRO336730050 ACRO336730250
Chlorodimethylvinylsilane 97%
Chlorodimethylvinylsilane
Vzorec: C₄H₉ClSi
Bod vzplanutí: –4 °C
MDL Number: MFCD00018090
CAS číslo: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4170 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR