Tisk…

Ethyl-2-amino-1-cyclohexene-1-carboxylate 98%

Dodavatel: Thermo Fisher Scientific

334260010
ACRO334260010EA 6660 CZK
ACRO334260010
Ethyl-2-amino-1-cyclohexene-1-carboxylate 98%
Ethyl-2-amino-1-cyclohexene-1-carboxylate

Appearance: White to light yellow Crystalline powder or crystals

Vzorec: C₉H₁₅NO₂
Bod tání: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01863243
CAS číslo: 1128-00-3

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 74 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR