Tisk…

Diethyl (chloromethyl)phosphonate 98%

Dodavatel: Thermo Fisher Scientific
Varování

330650010 330650050
ACRO330650010EA 1840 CZK
ACRO330650010 ACRO330650050
Diethyl (chloromethyl)phosphonate 98%
Diethyl (chloromethyl)phosphonate

Appearance: Clear colorless to light yellow Liquid

Vzorec: C₅H₁₂ClO₃P
Bod varu: 109…110 °C (10 mmHg)
Hustota: 1,2 g/cm³
Bod vzplanutí: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00010189
CAS číslo: 3167-63-3

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR