Tisk…

Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilised for molecular biology DNase-, RNase-, protease-free pH 7.8-8.2

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

327115000 327110025 327111000
ACRO327115000EA 11000 CZK
ACRO327115000 ACRO327110025 ACRO327111000
Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilised for molecular biology DNase-, RNase-, protease-free pH 7.8-8.2
Phenol : Chloroform : iso-Amyl alcohol

  • pH 7.8-8.2
  • saturated with 100 mM Tris-EDTA to pH 8.0

Hustota: 1,28 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00133763
CAS číslo: 136112-00-0
UN: 2810
ADR: 6.1,II

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to orange Solution
Stabilizer 0.08 to 0.12 % hydroxyquinoline
pH 7.8 to 8.2 (at 25°C)
Composition mixture of Phenol/Chloroform/Isoamyl alcohol in the ratio of 25:24:1 (v/v/v)
Dnase,Rnase,Protease activity None detected
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR