Tisk…

Methyl mercaptan sodium salt 95%, pure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Sodium methanethiolate, Sodium thiomethoxide, Sodium methylsulphanide, Methanethiol sodium salt

305710050 305710010 305710250 305711000
ACRO305710050EA 3790 CZK
ACRO305710050 ACRO305710010 ACRO305710250 ACRO305711000
Methyl mercaptan sodium salt 95%, pure
Methyl mercaptan sodium salt
Vzorec: CH₃NaS
MW: 70.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174316
CAS číslo: 5188-07-8
UN: 2921
ADR: 8 I

Objednat

Výsledky specifikačního testu

Appearance White to light beige Powder or granular powder
Titration Iodimetric 94 to 106 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR