Tisk…

Quinoline yellow sulphonic acid sodium salt (Acid yellow 3) 95%, water soluble, pure

Dodavatel: Thermo Fisher Scientific
Varování

277340250 277341000
ACRO277340250EA 4560 CZK
ACRO277340250 ACRO277341000
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3) 95%, water soluble, pure
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)

CAS No.: 8004-92-0

Vzorec: C₁₈H₁₁NO₂.2NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00080727
CAS číslo: 8004-92-0

Objednat

Výsledky specifikačního testu

Appearance Yellow to orange-brown Powder
Infrared spectrum Conforms
HPLC ≥94 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR