Tisk…

N-Boc-L-valine 99+%

Dodavatel: Thermo Fisher Scientific

275710050 275710250 275711000
ACRO275710050EA 784 CZK
ACRO275710050 ACRO275710250 ACRO275711000
N-Boc-L-valine 99+%
N-Boc-L-valine
Vzorec: C₁₀H₁₉NO₄
Bod tání: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065605
CAS číslo: 13734-41-3
EINECS: 237-307-6

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 76 to 80 °C
HPLC ≥99.0 % (ELSD)
Specific optical rotation -5.3° to -7.3° (20 °C, 589 nm) (c=1, CH3COOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR