Tisk…

N-Acetyl-β-D-glucosamine 98%

Dodavatel: Thermo Fisher Scientific

270940100 270941000 270945000
ACRO270940100EA 997 CZK
ACRO270940100 ACRO270941000 ACRO270945000
N-Acetyl-β-D-glucosamine 98%
N-Acetyl-β-D-glucosamine
Vzorec: C₈H₁₅NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00136044
CAS číslo: 7512-17-6
EINECS: 231-368-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (105°C, 4 hrs)
HPLC ≥97.5 % (ELSD)
Specific optical rotation +39° to +45° (20°C, 589 nm) (c=2, H2O, after 2 hrs)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR