Tisk…

Cadmium(II) sulphate 99+% for analysis

Dodavatel: Thermo Fisher Scientific
Nebezpečí

270565000 270560025
ACRO270565000EA 5450 CZK
ACRO270565000 ACRO270560025
Cadmium(II) sulphate 99+% for analysis
Cadmium(II) sulphate
Vzorec: CdO₄S
Bod tání: 1000 °C
Hustota: 3,79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010923
CAS číslo: 10124-36-4
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III
Merck Index: 13,01627

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Titration Complexometric ≥99.0 %
pH 3.5 to 6.0 (5 % at 20°C)
Chloride (Cl) ≤5 ppm
Arsenic (As) ≤0.5 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤100 ppm
Zinc (Zn) ≤50 ppm
Alkalies+earths (Na,K,Ca) ≤0.05 %
Nitrogen compounds (as N) ≤0.004 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR