Tisk…

Aceton 99.8% for HPLC

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

268310010 268310025
ACRO268310010EA 2390 CZK
ACRO268310010 ACRO268310025
Aceton 99.8% for HPLC
Aceton
Vzorec: CH₃COCH₃
Bod varu: 56,2 °C (1013 hPa)
Bod tání: –95 °C
Hustota: 0,79 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS číslo: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.8 %
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Residue after evaporation ≤0.0002 %
Acidity/alkalinity ≤0.0003 meq/g
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR