Tisk…

Sodium-tert-butanolate 99%, pure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate

252550050 252551000 252550025 252555000
ACRO252550050EA 1170 CZK
ACRO252550050 ACRO252551000 ACRO252550025 ACRO252555000
Sodium-tert-butanolate 99%, pure
Sodium-tert-butanolate
Vzorec: C₄H₉NaO
MW: 96.1
Bod tání: 180 °C
Bod vzplanutí: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00040575
CAS číslo: 865-48-5
EINECS: 212-741-9
UN: 3206
ADR: 4.2 II

Objednat

Výsledky specifikačního testu

Appearance White to light brown Powder
Titration with HCl 98.5 to 101.5 %
NaOH + Na2CO3 (as NaOH) ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR