Tisk…

L(-)-Carnitine hydrochloride 98%

Dodavatel: Thermo Fisher Scientific
Varování

230280050 230280250 230281000
ACRO230280050EA 1590 CZK
ACRO230280050 ACRO230280250 ACRO230281000
L(-)-Carnitine hydrochloride 98%
L(-)-Carnitine hydrochloride
Vzorec: C₇H₁₆ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00066100
CAS číslo: 6645-46-1
EINECS: 229-663-6

Objednat

Výsledky specifikačního testu

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Sulfated ash ≤0.5 %
Specific optical rotation -21° to -24° (20°C, 589 nm) (c=1, H2O)
Titration with NaOH 97.5 to 102.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR