Tisk…

Citrinin 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

229580050
ACRO229580050EA 8870 CZK
ACRO229580050
Citrinin 98%
Citrinin

Appearance: Yellow Crystalline powder or flakes

Vzorec: C₁₃H₁₄O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00006912
CAS číslo: 518-75-2
UN: 2811
ADR: 6.1,II

Objednat

Výsledky specifikačního testu

Appearance Yellow Crystalline powder or flakes
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR