Tisk…

Tetradecyltrimethylammonium bromide 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB

229370050 229371000 229375000 229370025
ACRO229370050EA 1020 CZK
ACRO229370050 ACRO229371000 ACRO229375000 ACRO229370025
Tetradecyltrimethylammonium bromide 99%
Tetradecyltrimethylammonium bromide
Vzorec: C₁₇H₃₈BrN
MW: 336.4
Bod tání: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011770
CAS číslo: 1119-97-7
EINECS: 214-291-9
UN: 1759
ADR: 8 II
Merck Index: 12,06419

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 245 to 250 °C
Titration Argentometric 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR