Tisk…

Methyl-ɑ-D-mannopyranoside 99+%

Dodavatel: Thermo Fisher Scientific

229251000 229250250 229255000
ACRO229251000EA 4750 CZK
ACRO229251000 ACRO229250250 ACRO229255000
Methyl-ɑ-D-mannopyranoside 99+%
Methyl-alpha-D-mannopyranoside

CAS No.: 617-04-9

Vzorec: C₇H₁₄O₆
Bod tání: 186…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00063262
CAS číslo: 617-04-9

Objednat

Výsledky specifikačního testu

Appearance White to off-white to light grey Powder
Infrared spectrum Conforms
Melting point 186 to 196 °C
Water ≤0.5 %
Sulfated ash ≤0.2 %
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +77° to +81° (20 °C, 589 nm) (c=10, water)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR