Tisk…

Toluen-α-sulfonylfluorid 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

215740010 215740050 215740100 215740500
ACRO215740010EA 1750 CZK
ACRO215740010 ACRO215740050 ACRO215740100 ACRO215740500
Toluen-α-sulfonylfluorid 99%
Toluen-α-sulfonylfluorid
Vzorec: C₇H₇FO₂S
MW: 174.2
Bod tání: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS číslo: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 II

Objednat

Výsledky specifikačního testu

Appearance White Needles and/or agglomerates
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR