Tisk…

Cacodylic acid sodium salt trihydrate 98%, pure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

214970250 214971000 214975000 214970100
ACRO214970250EA 2820 CZK
ACRO214970250 ACRO214971000 ACRO214975000 ACRO214970100
Cacodylic acid sodium salt trihydrate 98%, pure
Cacodylic acid sodium salt trihydrate
Vzorec: (CH₃)₂As(O)ONa·3H₂O
Bod tání: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00149079
CAS číslo: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: 6.1,II

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR