Tisk…

Diphenylphosphinic chloride 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Diphenylphosphinyl chloride

212960100 212960500
ACRO212960100EA 2160 CZK
ACRO212960100 ACRO212960500
Diphenylphosphinic chloride 98%
Diphenylphosphinic chloride
Vzorec: C₁₂H₁₀ClOP
MW: 236.63 g/mol
Bod varu: 222 °C (16 mmHg)
Bod vzplanutí: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00002077
CAS číslo: 1499-21-4
EINECS: 216-107-2
UN: 2920
ADR: 8 II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6080 to 1.6120 (20°C, 589 nm)
GC ≥97.5 %

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR