Tisk…

Xylenol orange tetrasodium salt, water soluble, pure

Dodavatel: Thermo Fisher Scientific

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

211220010 211220050 211220100 211220500
ACRO211220010EA 1680 CZK
ACRO211220010 ACRO211220050 ACRO211220100 ACRO211220500
Xylenol orange tetrasodium salt, water soluble, pure
Xylenol orange tetrasodium salt
Vzorec: C₃₁H₂₈N₂Na₄O₁₃S
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS číslo: 3618-43-7
EINECS: 222-805-8

Objednat

Výsledky specifikačního testu

Appearance Red-brown to dark brown Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10.0 % (110°C, 1 h)
UV Lambda max 575 to 585 nm (0.1N NaOH)
Suitability (for Zn titration) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR