Tisk…

Florisil® 60-100 mesh for column chromatography

Dodavatel: Thermo Fisher Scientific

205450010 205452500 205455000 205450050
ACRO205450010EA 12800 CZK
ACRO205450010 ACRO205452500 ACRO205455000 ACRO205450050
Florisil® 60-100 mesh for column chromatography
Magnesium silicate
Vzorec: MgO₃Si
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS číslo: 1343-88-0
EINECS: 215-681-1

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Particle size 6.9 % (140 mesh) typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR