Tisk…

L-allo-Threonine 99%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH

198541000 198542500
ACRO198541000EA 1800 CZK
ACRO198541000 ACRO198542500
L-allo-Threonine 99%
L-allo-Threonine
Vzorec: C₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00064268
CAS číslo: 28954-12-3
EINECS: 249-327-2

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation +8° to +10° (20°C, 589 nm) (c=2, water)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR