Tisk…

Poly(methyl methacrylate), M.W. ∼15000 (GPC)

Dodavatel: Thermo Fisher Scientific

190690500 190691000
ACRO190690500EA 4890 CZK
ACRO190690500 ACRO190691000
Poly(methyl methacrylate), M.W. ∼15000 (GPC)
Poly(methyl methacrylate)

Appearance: White Beads

Vzorec: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Bod vzplanutí: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00134349
CAS číslo: 9011-14-7

Objednat

Výsledky specifikačního testu

Appearance White Beads
Infrared spectrum Conforms
Loss on drying ≤3 %
Additional info bead size (typically): 10 to 100 micron


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR