Tisk…

Carmine alum lake of carminic acid, pure, high purity stain_biological

Dodavatel: Thermo Fisher Scientific

Synonyms: CI 75470

190200050 190200250 190201000
ACRO190200050EA 2400 CZK
ACRO190200050 ACRO190200250 ACRO190201000
Carmine alum lake of carminic acid, pure, high purity stain_biological
Carmine
Vzorec: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167028
CAS číslo: 1390-65-4
EINECS: 215-724-4

Objednat

Výsledky specifikačního testu

Appearance Dark red to purple Powder
Infrared spectrum Conforms
UV spectrum Complies
Assay ≥42.0 % (Carminic acid)
Loss on drying ≤20 %
Ash ≤12 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR