Tisk…

tert-Butylchlorodiphenylsilane 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane

187500100 187500500 187502500
ACRO187500100EA 1640 CZK
ACRO187500100 ACRO187500500 ACRO187502500
tert-Butylchlorodiphenylsilane 98%
tert-Butylchlorodiphenylsilane
Vzorec: C₁₆H₁₉ClSi
Bod varu: 135 °C (,7 mmHg)
Hustota: 1,06 g/cm³
Bod vzplanutí: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00000497
CAS číslo: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear to hazy colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5665 to 1.5685 (20°C, 589 nm)
GC ≥97.5 %
UV at 500 nm A: ≤0.02
pH 3.5 to 5.0 (5 % in water)
Titration with NaOH 99.4 to 100.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR