Tisk…

Dibutyltin oxide 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

179361000 179365000
ACRO179361000EA 1260 CZK
ACRO179361000 ACRO179365000
Dibutyltin oxide 98%
Dibutyltin oxide
Vzorec: C₈H₁₈OSn
MW: 248.94
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS číslo: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1 III

Objednat

Výsledky specifikačního testu

Appearance White Powder or granular powder
Infrared spectrum Conforms
Chloride (Cl) ≤0.2 %
Tin (Sn) ≥47 %
Volatile matter ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR