Tisk…

Polyethylene glycol, M.W. ≈ M.W. 600,000

Dodavatel: Thermo Fisher Scientific

Synonyms: Poly(ethylene oxide), Macrogol, PEO, PEG, Carbowax

178612500 178615000
ACRO178612500EA 3790 CZK
ACRO178612500 ACRO178615000
Polyethylene glycol, M.W. ≈ M.W. 600,000
Polyethylene glycol
Vzorec: H(OCH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS číslo: 25322-68-3

Objednat

Výsledky specifikačního testu

Appearance White Granular powder
Infrared spectrum Conforms
Loss on drying ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR