Tisk…

Diethylether 99+% Stabilised, extra_pure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilization: BHT

176830010 176830050 176830250
ACRO176830010EA 1800 CZK
ACRO176830010 ACRO176830050 ACRO176830250
Diethylether 99+% Stabilised, extra_pure
Diethylether
Vzorec: (CH₃CH₂)₂O
MW: 74.12
Bod varu: 34.6 °C (760 mmHg)
Bod tání: –116 °C
Hustota: 0.714 g/cm³
Bod vzplanutí: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS číslo: 60-29-7
UN: 1155
ADR: 3 I
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3510 to 1.3540 (20°C, 589 nm)
Water ≤0.1 % (Coulometric)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 5 to 7 ppm BHT
Relative density (20°C) 0.714 to 0.716
Residue after evaporation ≤0.005 %
Acidity ≤0.001 meq/g
Peroxides ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR