Tisk…

Chloroform-D1 99.6+ atom % D for NMR spectroscopy

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

174881000 174885000
ACRO174881000EA 2340 CZK
ACRO174881000 ACRO174885000
Chloroform-D1 99.6+ atom % D for NMR spectroscopy
Chloroform-D1
Vzorec: CDCl₃
Bod varu: 60,8 °C (1013 hPa)
Bod tání: –64 °C
Hustota: 1,5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS číslo: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.60 %
Residue after evaporation ≤0.01 %
Calcium (Ca) ≤1 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.5 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.5 ppm
Total phosphorus (P) ≤0.2 ppm
Total silicon (Si) ≤0.05 ppm
Total sulfur (S) ≤0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR