Tisk…

Bicine 99+% p.a.

Dodavatel: Thermo Fisher Scientific

172651000 172650010 172652500
ACRO172651000EA 3680 CZK
ACRO172651000 ACRO172650010 ACRO172652500
Bicine 99+% p.a.
Bicine
Vzorec: C₆H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS číslo: 150-25-4
EINECS: 205-755-1

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder or crystals or needles
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Sulfated ash ≤0.1 %
UV at 250 nm A: ≤0.05
pH 4.0 to 5.5 (10 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR