Tisk…

Trifluoromethanesulphonic acid 99%, extra pure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

169890010 169890100 169890500 169892500 169890011
ACRO169890010EA 1740 CZK
ACRO169890010 ACRO169890100 ACRO169890500 ACRO169892500 ACRO169890011
Trifluoromethanesulphonic acid 99%, extra pure
Trifluoromethanesulphonic acid
Vzorec: CHF₃O₃S
Bod varu: 162 °C (760 mmHg)
Bod tání: –40 °C
Hustota: 1,69 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007514
CAS číslo: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.3260 to 1.3300 (20°C, 589 nm)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR