Tisk…

Methyl myristate 99%

Dodavatel: Thermo Fisher Scientific

165010025 165011000 165015000
ACRO165010025EA 1280 CZK
ACRO165010025 ACRO165011000 ACRO165015000
Methyl myristate 99%
Methyl myristate
Vzorec: C₁₅H₃₀O₂
Bod varu: 323 °C (1013 hPa)
Hustota: 0,863 g/cm³
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS číslo: 124-10-7
EINECS: 204-680-1

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4380 (20°C, 589 nm)
Density (20/4°C) 0.853 to 0.873 g/ml
GC ≥98.5 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR