Tisk…

1,5-Difenylkarbazid 98% ACS

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Diphenylcarbazide, sym-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, 1,5-Diphenylcarbonohydrazide, N'',N'''-Diphenylcarbonohydrazide, sym. Diphenylcarbazide

158840250 158841000 158845000
ACRO158840250EA 1990 CZK
ACRO158840250 ACRO158841000 ACRO158845000
1,5-Difenylkarbazid 98% ACS
1,5-Difenylkarbazid
Vzorec: C₁₃H₁₄N₄O
MW: 242.28
Bod varu: 454
Bod tání: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00003013
CAS číslo: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance light grey to light purple to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 173 to 176 °C
Sulfated ash ≤0.05 %
Solubility (in aqueous acetone) Passes test
HPLC ≥97.5 %
Sensitivity (to chromate) Passes test

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR