Tisk…

N-Hydroxysuccinimide 98+%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

157270250 157271000 157272500 157270010
ACRO157270250EA 1420 CZK
ACRO157270250 ACRO157271000 ACRO157272500 ACRO157270010
N-Hydroxysuccinimide 98+%
N-Hydroxysuccinimide
Vzorec: C₄H₅NO₃
MW: 115.09
Bod varu: 363
Bod tání: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS číslo: 6066-82-6
EINECS: 228-001-3

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder or crystals (May have a tendency to cake)
Infrared spectrum Conforms
Water ≤2.0 % (K.F.)
Solubility The product may contain some hydroxysuccinimide:ammonia adduct, which is virtually insoluble in acetone
Titration with NaOH 98.0 to 101.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR