Tisk…

(±)-Pipecolinic acid 99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (±)-2-Piperidinecarboxylic acid, DL-Pipecolinic acid, H-DL-Homopro-OH, Pipecolinic Acid, Piperidine-2-carboxylic acid, Pipecolinic acid

131280250 131281000
ACRO131280250EA 2100 CZK
ACRO131280250 ACRO131281000
(±)-Pipecolinic acid 99%
(±)-Pipecolinic acid
Vzorec: C₆H₁₁NO₂
MW: 129.16
Bod tání: 268…278 °C
MDL Number: MFCD00064347
CAS číslo: 535-75-1
EINECS: 208-616-3

Objednat

Výsledky specifikačního testu

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 268 to 278 °C (Dec)
Water ≤1 % (K.F.)
Loss on drying ≤1 % (100 °C, 4 hrs)
Sulfated ash ≤0.5 %
Titration with HClO4 ≥98.5 %
UV (3 M in water)
pH 5.0 to 7.0 (1 M in water)
Dnase,Rnase,Protease activity None detected
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Zinc (Zn) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR