Tisk…

Diethylether 99+% Stabilised, pure

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilization: BHT

123990010 123990050 123990025 123990100 123990250
ACRO123990010EA 1930 CZK
ACRO123990010 ACRO123990050 ACRO123990025 ACRO123990100 ACRO123990250
Diethylether 99+% Stabilised, pure
Diethylether
Vzorec: (CH₃CH₂)₂O
MW: 74.12
Bod varu: 34.6 °C (760 mmHg)
Bod tání: –116 °C
Hustota: 0.714 g/cm³
Bod vzplanutí: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS číslo: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3510 to 1.3540 (20°C, 589 nm)
Water ≤0.2 % (Coulometric)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 5 to 10 ppm BHT
Residue after evaporation ≤0.002 %
Peroxides ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR