Tisk…

Diphenyliodonium chloride 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

117330100 117330500
ACRO117330100EA 4930 CZK
ACRO117330100 ACRO117330500
Diphenyliodonium chloride 97%
Diphenyliodonium chloride
Vzorec: C₁₂H₁₀ClI
Bod tání: 227…228 °C (subl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011909
CAS číslo: 1483-72-3
EINECS: 216-049-8
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
Chloride and Bromide ≤0.02 % (as Cl)
Iodide (I) passes test
Sulfate (SO4) ≤150 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR