Tisk…

Diethyl (cyanomethyl)phosphonate 96%

Dodavatel: Thermo Fisher Scientific
Varování

114270100 114270500 114272500
ACRO114270100EA 2450 CZK
ACRO114270100 ACRO114270500 ACRO114272500
Diethyl (cyanomethyl)phosphonate 96%
Diethyl (cyanomethyl)phosphonate
Vzorec: C₆H₁₂NO₃P
MW: 177.14 g/mol
Bod varu: 101…102 °C (.4 mmHg)
Hustota: 1.09 g/cm³
Bod vzplanutí: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001893
CAS číslo: 2537-48-6
EINECS: 219-806-0
UN: 3278
ADR: 6.1 III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4312 to 1.4332 (20°C, 589 nm)
GC ≥95 %
Additional info Cr2O3: 0.00005 %
Particle size 205 µm (D50) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR