Tisk…
Avantor® ACE® C18, HPLC Columns, 5 µm
Dodavatel: Avantor
ACE-121-2546EA 23820 CZK
ACE-121-2546 ACE-121-0202 ACE-121-0203 ACE-121-0246 ACE-121-0302 ACE-121-0346 ACE-121-0502 ACE-121-0503 ACE-121-0504 ACE-121-0510 ACE-121-0520 ACE-121-0530 ACE-121-0546 ACE-121-1002 ACE-121-1003 ACE-121-1004 ACE-121-1008 ACE-121-1010 ACE-121-1020 ACE-121-1030 ACE-121-1046 ACE-121-1203 ACE-121-1204 ACE-121-1220 ACE-121-1246 ACE-121-1502 ACE-121-1503 ACE-121-1504 ACE-121-1508 ACE-121-1510 ACE-121-1520 ACE-121-1530 ACE-121-1546 ACE-121-2502 ACE-121-2503 ACE-121-2504 ACE-121-2508 ACE-121-2510 ACE-121-2520 ACE-121-2530 ACE-121-3504 ACE-121-3546 ACE-121-7502 ACE-121-7520 ACE-121-7546 ACE-121-0103GD ACE-121-3004 ACE-121-0101GD ACE-121-0102GD ACE-121-0110GD ACE-121-7530 ACE-121-3002 ACE-121-3020 ACE-121-3030 ACE-121-3046 ACE-121-12003 ACE-121-25005 ACE-121-1000075 ACE-121-1539 ACE-121-1008E ACE-121-2546P ACE-121-3039 ACE-121-2004 ACE-121-2046 ACE1212546MVK ACE-121-0120GD ACE-121-2002 ACE-121-0104GD ACETACE-121-2539
Avantor® ACE® C18, HPLC Columns, 5 µm
Kolony chromatografické
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographicMore Product Information