Tisk…

Anti-S100 A4 Goat Polyclonal Antibody

Dodavatel: Abnova
PAB7142
ABNOPAB7142EA 14910 CZK
ABNOPAB7142
Anti-S100 A4 Goat Polyclonal Antibody
Protilátky
Goat polyclonal antibody raised against synthetic peptide of S100A4.

Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.03-0.1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-3 ?g/ml

Type: Primary
Antigen: S100 A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR