Tisk…

Anti-PAX5 Goat Polyclonal Antibody

Dodavatel: Abnova
PAB6220
ABNOPAB6220EA 14910 CZK
ABNOPAB6220
Anti-PAX5 Goat Polyclonal Antibody
Protilátky
Goat polyclonal antibody raised against synthetic peptide of PAX5.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: PAX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu PAX5
Název antigenu Paired box 5
Environmentally Preferable
Synonyma antigenu transcription factor PAX 5|B-cell lineage specific activator|B cell specific activator protein|paired box homeotic gene 5|paired box protein Pax-5|B-cell-specific transcription factor
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Goat
Identifikační číslo genu 5079
Izotyp
Western blot Yes
ELISA Yes
Imunogen A synthetic peptide corresponding to human PAX5.
Sekvence DLEKNYPTPRTSR-C
Formát Liquid
Čištění Antigen affinity purification
Křížová adsorbce No
Skladovací pufr Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
Skladovací teplota Store at –20 °C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR