Tisk…

Anti-TUBB3 Chicken Polyclonal Antibody

Dodavatel: Abnova
PAB29093
ABNOPAB29093EA 22210 CZK
ABNOPAB29093
Anti-TUBB3 Chicken Polyclonal Antibody
Protilátky
Chicken polyclonal antibody raised against recombinant Human TUBB3.

Recommended Dilutions: Immunocytochemistry: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: TUBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR