Tisk…

Anti-USB1 Goat Polyclonal Antibody

Dodavatel: Abnova
PAB24559
ABNOPAB24559EA 14910 CZK
ABNOPAB24559
Anti-USB1 Goat Polyclonal Antibody
Protilátky
Goat polyclonal antibody raised against synthetic peptide of C16orf57.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: USB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR